missing translation for 'onlineSavingsMsg'
Learn More

BTX™ AgilePulse™ USB Foot Switch Dual Pedal

Catalog No. BTX470420
Change view
Click to view available options
Connection Type:
Output Cable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
BTX470420 Output Cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTX470420 Supplier BTX™ Supplier No. BTX470420

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AgilePulse™ USB Foot Switch Dual Pedal allows hands-free operation.

  • Functions in place of the start button on the front of the generator
  • Ideal when conducting in vivo/in ovo gene delivery or nuclear transfer experiments when both hands are needed for sample manipulation
  • Compatible with most BTX™ generators

Specifications

Format Footswitch
For Use With (Application) In-Vivo Gene Delivery, in-Ovo Gene Delivery and Nuclear Transfer/Cloning
For Use With (Equipment) ECM™ 830, ECM™ 2001+, Legacy ECM™ 2001, AgilePulse™, and Gemini X2 Generators and MicroJect 1000 A Microinjection Systems
Product Line BTX Electroporation
Product Type Electroporation accessory
Description AgilePulse Foot Pedal
Display Touchscreen
Depth (English) 3.5 in.
Depth (Metric) 8.9 cm
Height (English) 2 in.
Height (Metric) 51 mm
Includes Foot Pedal
Length (English) Cable 10 ft.
Length (Metric) 3.1 m
Width (English) 4.5 in.
Width (Metric) 114 mm
Sterility Non-sterile
Connection Type Output Cable
Interface Footswitch
Pulse Form Constant, Linear, Non-linear
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.