missing translation for 'onlineSavingsMsg'
Learn More

BTX™ AgilePulse™ MAX Waveform Generator Only

Catalog No. BTX470201N
Change view
Click to view available options
Connection Type:
Chamber Cable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
BTX470201N Chamber Cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTX470201N Supplier BTX™ Supplier No. 470201N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AgilePulse™ MAX Waveform Generator Only provide an advanced electroporation solution for fast, efficient transfection of up to 10 mL of cell suspension.

Includes a user-friendly, programmable waveform generator with patented AgilePulse™ technology, BTX™ patent-pending large-volume electroporation chamber, and proprietary BTXPress™ Cytoporation Medium T, optimized for large volume electroporation. IQ/OQ Now available upon request.

  • Scale-up
    • Transfection protocols readily scale-up from standard laboratory cuvettes to large-volume transfection in the AgilePulse™ MAX system.
  • Maximal Efficiency with Cytoporation Medium
    • BTXPress™ Cytoporation Medium T used with the AgilePulse™ MAX system has been optimized for maximal efficiency with a number of cell lines, including K562, A20, HEK293 and CHO-KI.
    • Compatible with a large range of transfectants including DNA, RNA, siRNA, and olignonucleotides.
    • Can be directly diluted in complete growth medium for post-electroporation cell culture.
  • Simple User Interface
    • All controls are operated with the simple touch screen on the front panel.
    • Data is quickly retrieved by USB key and can be analyzed for detailed pulse characteristics including pulse and pulse current.
  • AgilePulse™ Advantage
    • Transfection efficiency and cell viability are enhanced by specialized, programmable electrical pulse waveforms, particularly important for larger polynucleotide delivery such as DNA plasmids.
    • The patented AgilePulse™ technology combines a unique sequence of short high-intensity pulses to porate cell membranes, followed by long low-intensity pulses to further drive transfectants into cells via electrophoresis, while maintaining cell viability.

Specifications

For Use With (Application) CRISPR Transfections, Transfect Cells Such as Bone Marrow to Produce or Replace a Missing Protein, Deliver siRNA to Suppress Gene Expression, Deliver Genes for Permanent Gene Correction, Load Cells with a Drug for Drug Delivery, Cancer Immunotherapy, Transfect Eukaryotic Cells for Protein Production in Bioreactors, Large-Scale Production of Replication-Deficient Viruses
For Use With (Equipment) BTXpress Cytoporation Medium T or Cytoporation Medium T4, large volume electroporation chamber
Frequency 5 kHz to 1 Hz
Product Line BTX Electroporation
Product Type Electroporator
Voltage 50 to 1000 V
Description AgilePulse MAX Waveform Generator Only
Display Touchscreen
Depth (English) 15.7 in.
Depth (Metric) 40 cm
Height (English) 7.9 in.
Height (Metric) 20 cm
Includes Generator Only
Operating Temperature (English) 50°F to 104°F
Pulse Length Range 0.200 to 1000 ms
Operating Temperature (Metric) 10°C to 40°C
Pulse Width Range 0.050 to 10 ms
Volume (Metric) Up to 10 mL
Sterility Non-sterile
Connection Type Chamber Cable
Interface Touch Screen Display, Footswitch
Pulse Form Square Wave
Show More Show Less