missing translation for 'onlineSavingsMsg'
Learn More

BTX™ AgilePulse™ MAX Chamber Holder for Large Volume 5 mL Chambers

AgilePulse™ MAX Chamber Holder for Large Volume 5 mL Chambers for use with AgilePulse™ MAX.

Supplier:  BTX™ 470202N

Catalog No. BTX470202N

Only null left
Explore available promotions

Description

Description

  • cuvette module plugs directly into generator
  • Holds one cuvette
  • Safety swing shield secures cuvette in holder
Specifications

Specifications

Large Volume Electroporation
Chamber Holder
AgilePulse MAX Chamber Holder for Large Volume 5 ml Chambers
Chamber Holder Only
BNC Cable
Square Wave
AgilePulse™ MAX, ECM™ 2001+
50 to 1000 V
Touchscreen
Non-sterile
Digital User
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions