missing translation for 'onlineSavingsMsg'
Learn More

BTX™ AgilePulse™ In Vivo ID (Intra-Dermal) System

Catalog No. BTX470400N
Change view
Click to view available options
Connection Type:
Electrode Connector Cable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
BTX470400N Electrode Connector Cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTX470400N Supplier BTX™ Supplier No. 470400N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AgilePulse™ In Vivo ID (Intra-Dermal) System provides maximum efficiency DNA and RNA delivery to skin, muscle, and tumor tissues.

  • Used for animal immunization, vaccine development and gene therapy, provides an intra-muscular, intra-dermal or intra-tumor electroporation solution to produce maximum transfection efficiency
  • Simple user interface
  • Pulse Agile Advantage
  • Uniform, reliable electroporation
  • Simple, Fast Treatment

Specifications

For Use With (Application) Vaccine Development, Intra-Dermal Gene Delivery, Intra-Tumor Gene Delivery
Frequency 5 kHz to 1 Hz
Product Line BTX Electroporation
Product Type Electroporation System
Voltage 50 to 1000 V
Description AgilePulse In Vivo ID (Intra-Dermal) System
Display Touchscreen
Depth (English) 15.7 in.
Depth (Metric) 40 cm
Height (English) 7.9 in.
Height (Metric) 20 cm
Includes AgilePulse™ ID Generator, Handle for Parallel Needle Array (47-0000), 4-needle Parallel Needle Array (47-0040 - 3 Pk.), 6-needle Parallel Needle Array (47-0050 - 3 Pk.) and ID Software
Operating Temperature (English) 50°F to 104°F
Pulse Length Range 0.200 to 1000 ms
Operating Temperature (Metric) 10°C to 40°C
Pulse Width Range 0.050 to 10 ms
Weight (English) 25 lb.
Weight (Metric) 11.3 kg
Width (English) 12.6 in.
Width (Metric) 32 cm
Sterility Non-sterile
Connection Type Electrode Connector Cable
Interface Touch Screen Display, Footswitch
Pulse Form Square Wave
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.