missing translation for 'onlineSavingsMsg'
Learn More

BTX™ AgilePulse™ In Vivo ID (Intra-Dermal) Generator Only

Catalog No. BTX470401N
Change view
Click to view available options
Connection Type:
Electrode Connector Cable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
BTX470401N Electrode Connector Cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTX470401N Supplier BTX™ Supplier No. 470401N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AgilePulse™ In Vivo ID (Intra-Dermal) Generator Only provides maximum efficiency DNA and RNA delivery to skin, muscle, and tumor tissues.

  • Used for animal immunization, vaccine development and gene therapy, provides an intra-muscular, intra-dermal or intra-tumor electroporation solution to produce maximum transfection efficiency
  • Simple user interface
  • Pulse Agile Advantage
  • Uniform, reliable electroporation
  • Simple, Fast Treatment

Specifications

For Use With (Application) Vaccine Development, Intra-Dermal Gene Delivery, Intra-Tumor Gene Delivery
For Use With (Equipment) In Vivo electroporation accessories
Frequency 5 kHz to 1 Hz
Product Line BTX Electroporation
Product Type Electroporation Generator
Voltage 50 to 1000 V
Description AgilePulse In Vivo ID (Intra-Dermal) Generator Only
Display Touchscreen
Depth (English) 15.7 in.
Depth (Metric) 40 cm
Height (English) 7.9 in.
Height (Metric) 20 cm
Includes Generator Only
Operating Temperature (English) 50°F to 104°F
Pulse Length Range 0.200 to 1000 ms
Operating Temperature (Metric) 10°C to 40°C
Pulse Width Range 0.050 to 10 ms
Weight (English) 25 lb.
Weight (Metric) 11.3 kg
Width (English) 12.6 in.
Width (Metric) 32 cm
Sterility Non-sterile
Connection Type Electrode Connector Cable
Interface Touch Screen Display, Footswitch
Pulse Form Square Wave
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.