missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Agarose medium EEO, for electrophoresis, (Mr = 0.16 - 0.19)
CAS: 9012-36-6
$185.94 - $2,114.05
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC400420250
|
Thermo Scientific Chemicals
400420250 |
25 g | Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC400421000
|
Thermo Scientific Chemicals
400421000 |
100 g | Glass Bottle |
Each for $517.08
|
|
||||
|
AC400425000
|
Thermo Scientific Chemicals
400425000 |
500 g | Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 9012-36-6 | |
| MFCD00081294 |
Specifications
| Fine Crystalline Powder | |
| 86.5°C to 89.5°C (1.5% gel) | |
| 0.16 to 0.19 | |
| >1000g/cm2 (1% gel) | |
| MFCD00081294 | |
| Electrophoresis |
| 9012-36-6 | |
| Cream to White | |
| 34.5° to 37.5°C (1.5% gel) | |
| Authentic | |
| Solubility in water: soluble in hot water. Other solubilities: insoluble in most common organic solvents | |
| Agarose medium EEO |
Safety and Handling
EINECSNumber : 232-731-8
TSCA : TSCA
RUO – Research Use Only