missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose medium EEO, for electrophoresis, (Mr = 0.16 - 0.19)

CAS: 9012-36-6

Supplier:  Thermo Scientific Chemicals 400421000

Quantity 100 g
Packaging Glass Bottle

 View more versions of this product

Catalog No. AC400421000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9012-36-6
MFCD00081294

Specifications

Agarose medium EEO
9012-36-6
Cream to White
0.16 to 0.19
>1000g/cm2 (1% gel)
Glass Bottle
Electrophoresis
Fine Crystalline Powder
86.5°C to 89.5°C (1.5% gel)
100 g
34.5° to 37.5°C (1.5% gel)
Authentic
MFCD00081294
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-731-8

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions