missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose, low gelling temperature, Molecular Biology Reagent

CAS: 9012-36-6

$407.38 - $1,305.71

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ6631914
View Documents Promotion Details
Thermo Scientific Chemicals
J6631914
25 g
Each for $407.38
Only null left
 
AAJ6631922
View Documents Promotion Details
Thermo Scientific Chemicals
J6631922
100 g
Each for $1,305.71
Only null left
 
Description

Description

Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it. Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it. Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.

Solubility
Soluble in water.

Notes
Gel strength (1.5% w/v):>500g/cm^2; Gelling temperature: <24-28°C; EEO: <0.12
Specifications

Chemical Identifiers

9012-36-6
MFCD00081294

Specifications

Powder
65°C
MFCD00081294
Agarose, low gelling temperature, Molecular Biology Reagent
9012-36-6
White
Soluble in water.
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-731-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only