missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose, low gelling temperature, Molecular Biology Reagent

Catalog No. AAJ6631914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6631914 25 g
AAJ6631922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6631914 Supplier Thermo Scientific Chemicals Supplier No. J6631914

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9012-36-6

Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it. Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it. Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.

Solubility
Soluble in water.

Notes
Gel strength (1.5% w/v):>500g/cm^2; Gelling temperature: <24-28°C; EEO: <0.12

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Powder
Melting Point 65°C
Color White
Quantity 25 g
Solubility Information Soluble in water.
Chemical Name or Material Agarose, low gelling temperature, Molecular Biology Reagent
EINECSNumber 232-731-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only