missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose LM, low melt, for recovery of samples after separation

Catalog No. AAH2641722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2641722 100 g
AAH2641706 5 g
AAH2641714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2641722 Supplier Thermo Scientific Chemicals Supplier No. H2641722

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for size-based separation of nucleic acids in agarose gel electrophoresis applications. | CAS: 9012-36-6 | C24H38O19 | 630.55 g/mol

Used for size-based separation of nucleic acids in agarose gel electrophoresis applications. Ideal for analysis and recovery of DNA and RNA for routine applications. Ideal for a variety of applications such as in-gel manipulation, tissue and cell culture, and viral plaque assays. Can also be used in protein electrophoresis applications such as Ouchterlony (antigen-antibody interaction assay) and radial immunodiffusion (RID) (antigen quantitation assay)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9012-36-6

Specifications

Quantity 100 g
Sensitivity Hygroscopic; Moisture sensitive
Product Type LM Agarose

RUO – Research Use Only