missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose medium-low EEO, for electrophoresis, (Mr = 0.12)

CAS: 9012-36-6

$156.18 - $1,411.68

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC400410250
View Documents Promotion Details
Thermo Scientific Chemicals
400410250
25 g Glass Bottle
Each for $156.18
Only null left
 
AC400411000
View Documents Promotion Details
Thermo Scientific Chemicals
400411000
100 g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC400415000
View Documents Promotion Details
Thermo Scientific Chemicals
400415000
500 g Glass Bottle
Each for $1,411.68
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9012-36-6
MFCD00081294

Specifications

Fine Crystalline Powder
86.5°C to 89.5°C (1.5% gel)
0.05 to 0.13
>1200g/cm2 (1% gel), >2500g/cm2 (1.5% gel)
MFCD00081294
Agarose medium-low EEO
9012-36-6
Cream to White
34.5° to 37.5°C (1.5% gel)
Authentic
Electrophoresis
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-731-8

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only