missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose medium-low EEO, for electrophoresis, (Mr = 0.12)

Catalog No. AC400415000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC400415000 500 g Glass Bottle
AC400410250 25 g Glass Bottle
AC400411000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC400415000 Supplier Thermo Scientific Chemicals Supplier No. 400415000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9012-36-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Fine Crystalline Powder
Melting Point 86.5°C to 89.5°C (1.5% gel)
Color Cream to White
Quantity 500 g
Electroendosmosis (EEO) 0.05 to 0.13
Gelation Temperature 34.5° to 37.5°C (1.5% gel)
Gel Strength >1200g/cm2 (1% gel), >2500g/cm2 (1.5% gel)
Infrared Spectrum Authentic
Packaging Glass Bottle
Grade Electrophoresis
Chemical Name or Material Agarose medium-low EEO
Show More Show Less
EINECSNumber 232-731-8
TSCA TSCA

RUO – Research Use Only