missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose, high melting temperature, medium resolution

For standard DNA applications | CAS: 9012-36-6

Supplier:  Thermo Scientific Chemicals J6112314

 View more versions of this product

Catalog No. AAJ6112314


Only null left
Explore available promotions

Description

Description

For standard DNA applications.Agarose, High Melting Temperature, Medium Resolution is used for standard DNA separations. It is involved in the purification of protein by using column-based preparative scale separation as in gel filtration chromatography and ion exchange chromatography

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
For standard DNA applications.Agarose, High Melting Temperature, Medium Resolution is used for standard DNA separations. It is involved in the purification of protein by using column-based preparative scale separation as in gel filtration chromatography and ion exchange chromatography

Solubility
Soluble in water.

Notes
Melting temp: ∼88°.Moisture sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Specifications

Specifications

Agarose
Powder
White
MFCD00081294
high melting temperature, medium resolution
9012-36-6
25 g
Soluble in water.

Chemical Identifiers

9012-36-6
MFCD00081294
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-731-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only