missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose, high melting temperature, medium resolution

For standard DNA applications | CAS: 9012-36-6

$188.03 - $1,483.30

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ6112314
View Documents Promotion Details
Thermo Scientific Chemicals
J6112314
25 g
Each for $188.03
Only null left
 
AAJ6112322
View Documents Promotion Details
Thermo Scientific Chemicals
J6112322
100 g
Each for $478.77
Only null left
 
AAJ6112336
View Documents Promotion Details
Thermo Scientific Chemicals
J6112336
500 g
Each for $1,483.30
Only null left
 
Description

Description

For standard DNA applications.Agarose, High Melting Temperature, Medium Resolution is used for standard DNA separations. It is involved in the purification of protein by using column-based preparative scale separation as in gel filtration chromatography and ion exchange chromatography

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
For standard DNA applications.Agarose, High Melting Temperature, Medium Resolution is used for standard DNA separations. It is involved in the purification of protein by using column-based preparative scale separation as in gel filtration chromatography and ion exchange chromatography

Solubility
Soluble in water.

Notes
Melting temp: ∼88°.Moisture sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Specifications

Chemical Identifiers

9012-36-6
MFCD00081294

Specifications

Powder
White
Soluble in water.
9012-36-6
MFCD00081294
Agarose, high melting temperature, medium resolution
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-731-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only