missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose, high gelling temperature, Molecular Biology Reagent

CAS: 9012-36-6

$322.08 - $625.00

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ6670414
View Documents Promotion Details
Thermo Scientific Chemicals
J6670414
25 g
Each for $322.08
Only null left
 
AAJ6670422
View Documents Promotion Details
Thermo Scientific Chemicals
J6670422
100 g
Each for $625.00
Only null left
 
Description

Description

High gel strength allows gel formation at low concentrations. Ideal for antibiotic sensitivity assays, RID, double diffusion and other immunological applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • High gel strength allows gel formation at low concentrations
  • Ideal for antibiotic sensitivity assays, RID, double diffusion and other immunological applications
Specifications

Chemical Identifiers

9012-36-6
MFCD00081294

Specifications

Powder
White
MFCD00081294
9012-36-6
Hygroscopic; Moisture sensitive
Agarose, high gelling temperature, Molecular Biology Reagent
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-731-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only