missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Agarase (0.5 U/μL)

Catalog No. FEREO0461
Change view
Click to view available options
Quantity:
100 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREO0461 100 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEREO0461 Supplier Thermo Scientific™ Supplier No. EO0461
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifically digests the agarose polysaccharide core, which allows for gentle yet efficient recovery of DNA or RNA fragments from low melting point agarose.

Agarase specifically digests the agarose polysaccharide core into neoagaro-oligosaccharides. Agarase allows for gentle yet efficient recovery of DNA or RNA fragments from low melting point agarose. The recovered nucleic acids can be directly used for amplification, cloning, sequencing, etc.

Highlights

  • Gentle -recovers intact DNA and RNA
  • High recovery yields -> 90% of the starting DNA recovered following extraction
  • Ideal for both long (> 30 kb) and short (< 100 bp) DNA fragments 
  • Active in TAE and TBE electrophoresis buffers 

Specifications

Chemical Name or Material Agarose
Concentration 0.5 U/μL
For Use With (Application) Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), PCR, Cloning, cDNA Library Construction, Sequencing, In Vitro Transcription, Nuclease Protection Assays, Nucleic Acid Labeling
Quantity 100 units

For Research Use Only. Not for use in diagnostic procedures.