missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Agar -Y Powder
Use for yeast cloning growth and spheroplast regeneration
$545.21 - $545.21
Specifications
| Content And Storage | 15°C to 30°C |
|---|---|
| Additive | Agar -Y |
| Form | Powder |
| Grade | Superior |
| For Use With (Application) | Yeast cell culture |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
MP114019032
|
MP Biomedicals™
114019032 |
1000 g |
Each for $545.21
|
|
|||||
Description
Yeast cell culture
Specifications
| 15°C to 30°C | |
| Powder | |
| Yeast cell culture |
| Agar -Y | |
| Superior |