missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Agar -Y Powder
Description
Yeast cell culture
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Additive | Agar -Y |
| Form | Powder |
| Grade | Superior |
| For Use With (Application) | Yeast cell culture |
| Quantity | 1000 g |