Learn More
Thermo Scientific Chemicals Agar powder
CAS: 9002-18-0 | C14H24O9 | 336.337 g/mol
Supplier: Thermo Scientific Chemicals A107520E
Description
Agar is the most suited solidifying agent for scientific applications that require incubation temperatures closer to human body temperature. Agar is used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAgar is the most suited solidifying agent for scientific applications that require incubation temperatures closer to human body temperature. Agar is used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.
Solubility
Soluble in water at temperatures between 950C and 1000 C. Insoluble in cold water.
Notes
Prolonged exposure to high temperatures can degrade Agar powder.
Specifications
| Powder | |
| 9002-18-0 | |
| MFCD00081288 | |
| agar, agar, pure, powder, agar agar bacteriological, 3r,4s,5s,6r-2-4r,5s-4-hydroxy-3-methyl-2,6-dioxabicyclo 3.2.1 octan-8-yl oxy-6-hydroxymethyl-4-methoxyoxane-3,5-diol | |
| GYYDPBCUIJTIBM-DYOGSRDZSA-N | |
| (2R,3S,4S,5R)-2-(hydroxymethyl)-6-[[(4R,5S)-4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy]-4-methoxyoxane-3,5-diol | |
| 71571511 |
| Agar | |
| C14H24O9 | |
| 2500 g | |
| Soluble in water at temperatures between 950C and 1000C. Insoluble in cold water. | |
| CC1C(C2C(C(O1)CO2)OC3C(C(C(C(O3)CO)O)OC)O)O | |
| 336.337 |
Chemical Identifiers
| 9002-18-0 | |
| 336.337 | |
| GYYDPBCUIJTIBM-DYOGSRDZSA-N | |
| 71571511 | |
| CC1C(C2C(C(O1)CO2)OC3C(C(C(C(O3)CO)O)OC)O)O |
| C14H24O9 | |
| MFCD00081288 | |
| agar, agar, pure, powder, agar agar bacteriological, 3r,4s,5s,6r-2-4r,5s-4-hydroxy-3-methyl-2,6-dioxabicyclo 3.2.1 octan-8-yl oxy-6-hydroxymethyl-4-methoxyoxane-3,5-diol | |
| (2R,3S,4S,5R)-2-(hydroxymethyl)-6-[[(4R,5S)-4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy]-4-methoxyoxane-3,5-diol |
Safety and Handling
EINECSNumber : 232-658-1
RTECSNumber : AW7950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only