missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agar powder

Catalog No. p-4408666
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1075222 100 g
AAA1075236 500 g
AAA107520E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1075222 Supplier Thermo Scientific Chemicals Supplier No. A1075222
Only null left

CAS: 9002-18-0 | C14H24O9 | 336.337 g/mol

Agar is the most suited solidifying agent for scientific applications that require incubation temperatures closer to human body temperature. Agar is used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agar is the most suited solidifying agent for scientific applications that require incubation temperatures closer to human body temperature. Agar is used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.

Solubility
Soluble in water at temperatures between 950C and 1000 C. Insoluble in cold water.

Notes
Prolonged exposure to high temperatures can degrade Agar powder.

Chemical Identifiers

CAS 9002-18-0
Molecular Formula C14H24O9
Molecular Weight (g/mol) 336.337
MDL Number MFCD00081288
InChI Key GYYDPBCUIJTIBM-DYOGSRDZSA-N
Synonym agar, agar, pure, powder, agar agar bacteriological, 3r,4s,5s,6r-2-4r,5s-4-hydroxy-3-methyl-2,6-dioxabicyclo 3.2.1 octan-8-yl oxy-6-hydroxymethyl-4-methoxyoxane-3,5-diol
PubChem CID 71571511
IUPAC Name (2R,3S,4S,5R)-2-(hydroxymethyl)-6-[[(4R,5S)-4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy]-4-methoxyoxane-3,5-diol
SMILES CC1C(C2C(C(O1)CO2)OC3C(C(C(C(O3)CO)O)OC)O)O

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Soluble in water at temperatures between 950C and 1000C. Insoluble in cold water.
Chemical Name or Material Agar
EINECSNumber 232-658-1
RTECSNumber AW7950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only