missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Agar, NF Grade
Powder, meets N.F. grade
Supplier: MP Biomedicals™ 0210026280
Description
- 80-100 mesh
- Substitute for gelatin, isinglass
- Used as nutrient media for bacterial culture
Specifications
| 15°C to 30°C | |
| USP, NF | |
| Used for nutrient media for bacterial cultures |
| Powder | |
| Agar | |
| 100 g |