missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Agar, NF Grade

Powder, meets N.F. grade

$115.76 - $412.92

Specifications

Content And Storage 15°C to 30°C
Form Powder
Grade USP, NF
Product Type Agar
For Use With (Application) Used for nutrient media for bacterial cultures
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10026283
View Documents
MP Biomedicals™
0210026283
250 g
Each for $238.30
Only null left
 
ICN10026280
View Documents
MP Biomedicals™
0210026280
100 g
Each for $115.76
Only null left
 
ICN10026290
View Documents
MP Biomedicals™
0210026290
500 g
Each for $412.92
Only null left
 
Description

Description

  • 80-100 mesh
  • Substitute for gelatin, isinglass
  • Used as nutrient media for bacterial culture
Specifications

Specifications

15°C to 30°C
USP, NF
Used for nutrient media for bacterial cultures
Powder
Agar
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'