Learn More
MP Biomedicals™ Agar, NF Grade
Powder, meets N.F. grade
Supplier:  MP Biomedicals™ 0210026290
 View more versions of this product
Catalog No. ICN10026290
Only null left
Explore available promotions
Description
Description
  • 80-100 mesh
  • Substitute for gelatin, isinglass
  • Used as nutrient media for bacterial culture
Specifications
Specifications
15°C to 30°C
USP, NF
Used for nutrient media for bacterial cultures
Powder
Agar
500 g
Product Suggestions
Product Suggestions