missing translation for 'onlineSavingsMsg'
Learn More

Aflatoxin B1-13C17 Solution, 0.5 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111017924
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017924 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017924 Supplier Merck Emd Millipore Supplier No. 327641ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Aflatoxin B1-13C17 is the 13C isotope-labeled analog of the mycotoxin, aflatoxin.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 1217449-45-0
Linear Formula 13C17H12O6
MDL Number MFCD16628323
Synonym Fully 13 C-labelled Aflatoxin B1
Type Analytical Standard
Quantity 1 mL
Formula Weight 329.15
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 0.5 μg/mL in acetonitrile