missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AF9 (MLLT3) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE259725UG
Description
AF9 (MLLT3) Polyclonal antibody specifically detects AF9 (MLLT3) in Human, Mouse samples. It is validated for ChIP-Seq, Immunocytochemistry, Western Blotting.Chemical Identifiers
| AF9 (MLLT3) | |
| ChIP sequencing (ChIP-seq), Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminal region of human AF9 (MLLT3) protein. | |
| Primary | |
| Stable for 1 year at 2-8°C from date of receipt. |
| 1.0 mg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| IgG | |
| Affinity Purified |
Specifications
| AF9 (MLLT3) | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Human, Mouse | |
| Purified |
| ChIP sequencing (ChIP-seq), Immunocytochemistry, Western Blot | |
| 1.0 mg/mL | |
| Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminal region of human AF9 (MLLT3) protein. | |
| 25 μg | |
| Primary | |
| Stable for 1 year at 2-8°C from date of receipt. | |
| IgG |