missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals AEBSF hydrochloride, 98%

CAS: 30827-99-7 | C8H10FNO2S·ClH | 239.7 g/mol

Supplier:  Thermo Scientific Chemicals 328110500

 View more versions of this product

Catalog No. AC328110500


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

AEBSF hydrochloride
30827-99-7
100.0
White
98%
C8H10FNO2S·ClH
50 mg
Solubility in water: 10g/L (20°C). Other solubilities: soluble in absolute ethanol
C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl
239.7
239.7
Powder
98%
97.5
175.0°C to 177.0°C
Authentic
Glass bottle
MFCD00132962
4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride
WRDABNWSWOHGMS-UHFFFAOYSA-N
4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride
186136
98%

Chemical Identifiers

30827-99-7
239.7
WRDABNWSWOHGMS-UHFFFAOYSA-N
186136
C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl
C8H10FNO2S·ClH
MFCD00132962
4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride
4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only