missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Advanced TC Cell Culture Dishes
GREENER_CHOICE

Catalog No. 07000686
Change view
Click to view available options
Diameter (Metric):
100 mm
145 mm
Volume (Metric) Working:
15 to 17 mL
25 to 27 mL
Height (Metric):
20 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Volume (Metric) Working Height (Metric)
07000686 100 mm 15 to 17 mL 20 mm
07000598 145 mm 25 to 27 mL 20 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07000686 Supplier Greiner Bio-One Supplier No. 664960

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For cultivation of fastidious and sensitive cells. Greiner Bio-One Advanced TC Cell Culture Dishes are free of detectable DNase, RNase, human DNA.

  • Non-pyrogenic and non-cytotoxic
  • Optimised cell adherence
  • Increased cell yield
  • Homogenous cell growth
  • Higher transgene activity
  • Improved assay consistency
  • Non-biological surface, sterile
  • Shelf

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Flat
Capacity (Metric) 100 mL
Color Clear
Culture Area 58 sq. cm
Material Polystyrene
Shape Round
Sterility Sterile
Surface Treatment Advanced TC
Type Cell Culture Dish
Volume (Metric) Working 15 to 17 mL
Includes Lid
Height (Metric) 20 mm
Diameter (Metric) 100 mm
Show More Show Less