Learn More
United Scientific™ Advanced Glassware Set of 9
Advanced Glassware Set of 9 is safely packed to ensure break free storage.
Supplier:  United Scientific™ EGSSET9
Catalog No. S03091
Only null left
Explore available promotions
Description
Description
An ideal starter set for any laboratory, this set contains low form beakers, Erlenmeyer flasks, and Class A graduated cylinders. Borosilicate glass is designed to give the best combination of thermal shock resistance and physical strength.
  • Features heavy-duty rims and graduations in durable white enamel. Includes reusable storage box with foam insert
  • Kit Contains: Low Form Glass Beaker Set - 3 sizes - 50 mL, 100 mL and 250 mL (White Graduations)
  • Glass Graduated Cylinder Set - 3 Sizes - 10 mL, 50 mL and 100 mL (Amber Graduations, To Contain)
  • Glass Erlenmeyer Flask Set - 3 Sizes - 50 mL, 150 mL and 250 mL (White Graduations)
Specifications
Specifications
Borosilicate Glass
Set of 9
Chemistry Kit
Product Suggestions
Product Suggestions