missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADORA2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572850
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572850 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572850 Supplier Invitrogen™ Supplier No. PA572850
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

ADORA2B Polyclonal Antibody for Western Blot, IHC (P), ELISA

ADORA2B encodes an adenosine receptor that is a member of the G protein-coupled receptor superfamily. This integral membrane protein stimulates adenylate cyclase activity in the presence of adenosine. This protein also interacts with netrin-1, which is involved in axon elongation. The gene is located near the Smith-Magenis syndrome region on chromosome 17.

Specifications

Antigen ADORA2B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ADORA2B
Gene Accession No. P29275, P29276, Q60614
Gene Alias A2b; A2b receptor; A2b, Rs; A2BAR; A2BR; AA2BR; adenosine A2B receptor; adenosine receptor A2b; ADORA2; Adora2b; AI480866; AI605384; HGNC:264; netrin-1 receptor
Gene Symbols ADORA2B
Host Species Rabbit
Immunogen A 19 amino acid peptide near the carboxy terminus of human ADORA2B, located within the last 50 amino acids.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11541, 136, 29316
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less