missing translation for 'onlineSavingsMsg'
Learn More

Interscience Adjustable Plate for Petri Dish

Catalog No. 16948979
Change view
Click to view available options
Holds:
Petri Dishes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
16948979 Petri Dishes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16948979 Supplier Interscience Supplier No. 413019
Only null left

Adjustable plate for Petri dish

  • Multiadaptable support for Petri dishes
  • Adjustable from 90 to 150 mm

Specifications

Color Silver
Holds Petri Dishes
Shape Round
For Use With (Equipment) EasySpiral
Product Type Petri Dish Holder
Diameter (Metric) Inner 90 to 150 mm