missing translation for 'onlineSavingsMsg'
Learn More

Adipoyl Chloride 98.0+%, TCI America™

Supplier:  TCI America A0169100G

 View more versions of this product

Catalog No. A0169100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Adipoyl Chloride
Yellow
C6H8Cl2O2
100 g
adipoyl chloride, adipoyl dichloride, adipyl chloride, adipoylchloride, adipic dichloride, hexanedioyl chloride, adipic acid dichloride, butane-1,4-dicarbonyl chloride, adipic acid chloride, pubchem13242
C(CCC(=O)Cl)CC(=O)Cl
183.028
183.03
Liquid
111-50-2
115°C
MFCD00000759
3265
PWAXUOGZOSVGBO-UHFFFAOYSA-N
hexanedioyl dichloride
61034
≥98.0% (GC,T)

Chemical Identifiers

111-50-2
183.028
PWAXUOGZOSVGBO-UHFFFAOYSA-N
61034
C(CCC(=O)Cl)CC(=O)Cl
C6H8Cl2O2
MFCD00000759
adipoyl chloride, adipoyl dichloride, adipyl chloride, adipoylchloride, adipic dichloride, hexanedioyl chloride, adipic acid dichloride, butane-1,4-dicarbonyl chloride, adipic acid chloride, pubchem13242
hexanedioyl dichloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2563

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions