missing translation for 'onlineSavingsMsg'
Learn More

Adipoyl chloride, 98%

Catalog No. p-50939
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173181000 100 mL Glass bottle
AC173185000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC173181000 Supplier Thermo Scientific Chemicals Supplier No. 173181000
Only null left

Adipoyl chloride, 98%, C6H8Cl2O2, CAS Number-111-50-2, adipic acid chloride, adipic acid dichloride, adipic dichloride, adipoylchloride, adipoyl chloride, adipoyl dichloride, adipyl chloride, butane-1,4-dicarbonyl chloride, hexanedioyl chloride, pubchem13242, 500mL, 02, 653 | CAS: 111-50-2 | C6H8Cl2O2 | 183.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-50-2
Molecular Formula C6H8Cl2O2
Molecular Weight (g/mol) 183.03
MDL Number MFCD00000759
InChI Key PWAXUOGZOSVGBO-UHFFFAOYSA-N
Synonym adipoyl chloride, adipoyl dichloride, adipyl chloride, adipoylchloride, adipic dichloride, hexanedioyl chloride, adipic acid dichloride, butane-1,4-dicarbonyl chloride, adipic acid chloride, pubchem13242
PubChem CID 61034
IUPAC Name hexanedioyl dichloride
SMILES C(CCC(=O)Cl)CC(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown
Density 1.2500g/mL
Boiling Point 105.0°C to 107.0°C (2.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4700 to 1.4720
Linear Formula ClCO(CH2)4COCl
Quantity 100 mL
Beilstein 02, 653
Specific Gravity 1.25
Solubility Information Solubility in water: reacts. Other solubilities: forming hcl
Formula Weight 183.03
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Adipoyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Contact with water liberates toxic gas.
Reacts violentl
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 203-876-4
TSCA TSCA

RUO – Research Use Only