missing translation for 'onlineSavingsMsg'
Learn More

Adiponitrile 98.0+%, TCI America™

Catalog No. p-7116752
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A016825ML 25 mL
A0168500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A016825ML Supplier TCI America Supplier No. A016825ML
Only null left

Chemical Identifiers

CAS 111-69-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00001975
InChI Key BTGRAWJCKBQKAO-UHFFFAOYSA-N
Synonym adiponitrile, 1,4-dicyanobutane, adipodinitrile, adipyldinitrile, adipic acid dinitrile, tetramethylene dicyanide, nitrile adipico, tetramethylene cyanide, adipic acid nitrile, adipinsaeuredinitril
PubChem CID 8128
IUPAC Name hexanedinitrile
SMILES C(CCC#N)CC#N

Specifications

Melting Point 2°C
Color Yellow
Boiling Point 295°C
Quantity 25 mL
UN Number 2205
Formula Weight 108.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Adiponitrile
EINECSNumber (2)-1512
RTECSNumber AV2625000
TSCA Yes