missing translation for 'onlineSavingsMsg'
Learn More

Adiponitrile 98,0+%, TCI America™

Catalog No. A0168500ML
Change view
Click to view available options
Quantité:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
A0168500ML 500 mL
A016825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0168500ML Supplier TCI America Supplier No. A0168500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 111-69-3
Formule moléculaire C6H8N2
Poids moléculaire (g/mol) 108.144
Numéro MDL MFCD00001975
Clé InChI BTGRAWJCKBQKAO-UHFFFAOYSA-N
Synonyme adiponitrile, 1,4-dicyanobutane, adipodinitrile, adipyldinitrile, adipic acid dinitrile, tetramethylene dicyanide, nitrile adipico, tetramethylene cyanide, adipic acid nitrile, adipinsaeuredinitril
PubChem CID 8128
Nom de l’IUPAC Hexanedinitrile
SOURIRES C(CCC#N)CC#N

Specifications

Nom ou substance chimique Adiponitrile
Point de fusion 2 °C
Couleur Jaune
Point d’ébullition 295 °C
Quantité 500 mL
Numéro UN 2205
Poids de formule 108.14
Pourcentage de pureté ≥98,0% (GC)
Forme physique Liquide
EINECSNumber (2)-1512
RTECSNumber AV2625000
TSCA Oui