missing translation for 'onlineSavingsMsg'
Learn More

Adiponitrile, 99+%, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. p-7058964
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43679AE 100 mL
AA43679AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43679AE Supplier Thermo Scientific Chemicals Supplier No. 043679AE
Only null left

CAS: 111-69-3 | C6H8N2 | 108.144 g/mol

Adiponitrile is used as an intermediate for the synthesis of hexamethylenediamine (diamino-1,6-hexane). This chemical is an important intermediate for the manufacture of synthetic fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon

Chemical Identifiers

CAS 111-69-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00001975
InChI Key BTGRAWJCKBQKAO-UHFFFAOYSA-N
Synonym adiponitrile, 1,4-dicyanobutane, adipodinitrile, adipyldinitrile, adipic acid dinitrile, tetramethylene dicyanide, nitrile adipico, tetramethylene cyanide, adipic acid nitrile, adipinsaeuredinitril
PubChem CID 8128
IUPAC Name hexanedinitrile
SMILES C(CCC#N)CC#N

Specifications

Melting Point 2°C
Density 0.951 g/mL
Boiling Point 295°C
Flash Point 163°C (325°F)
Odor Odorless
Packaging ChemSeal™ bottles
Refractive Index 1.438
Quantity 100 mL
UN Number UN2205
Beilstein 1740005
Solubility Information Highly soluble in water (83g/L).
Formula Weight 108.14
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Adiponitrile
Show More Show Less
Hazard Category H301-H332
Hazard Statement GHS H Statement
H301-H331-H227
Toxic if swallowed.
Toxic if inhaled.
Combustible liquid.
Precautionary Statement P261-P264b-P270-P271-P301+P310-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ADIPONITRILE
EINECSNumber 203-896-3
RTECSNumber AV2625000
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures

RUO – Research Use Only