missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adipic Dihydrazide 99.0+%, TCI America™
Supplier: TCI America A0170250G
Specifications
| Adipic Dihydrazide | |
| 1071-93-8 | |
| C6H14N4O2 | |
| adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5m | |
| NNC(=O)CCCCC(=O)NN | |
| 174.20 | |
| 174.20 | |
| 250 g |
| 182°C | |
| White | |
| MFCD00007614 | |
| IBVAQQYNSHJXBV-UHFFFAOYSA-N | |
| hexanedihydrazide | |
| 66117 | |
| ≥99.0% (HPLC) | |
| Crystalline Powder |
Chemical Identifiers
| 1071-93-8 | |
| 174.20 | |
| IBVAQQYNSHJXBV-UHFFFAOYSA-N | |
| 66117 | |
| NNC(=O)CCCCC(=O)NN |
| C6H14N4O2 | |
| MFCD00007614 | |
| adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5m | |
| hexanedihydrazide |
Safety and Handling
EINECSNumber : (2)-0865
RTECSNumber : AV1400000
TSCA : Yes