missing translation for 'onlineSavingsMsg'
Learn More

Adipic Dihydrazide 99.0+%, TCI America™

Catalog No. p-7116754
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A017025G 25 g
A0170250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A017025G Supplier TCI America Supplier No. A017025G
Only null left

Chemical Identifiers

CAS 1071-93-8
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00007614
InChI Key IBVAQQYNSHJXBV-UHFFFAOYSA-N
Synonym adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5m
PubChem CID 66117
IUPAC Name hexanedihydrazide
SMILES NNC(=O)CCCCC(=O)NN

Specifications

Melting Point 182°C
Color White
Formula Weight 174.20
Percent Purity ≥99.0% (HPLC)
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Adipic Dihydrazide
EINECSNumber (2)-0865
RTECSNumber AV1400000
TSCA Yes