missing translation for 'onlineSavingsMsg'
Learn More

Adipic Acid, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019091
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019091 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019091 Supplier Merck Emd Millipore Supplier No. 89143100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 124-04-9
Product Line TraceCERT
Vapor Pressure 1 mmHg (159.5°C)
Vapor Density 5 (vs. air)
Physical Form Neat
Linear Formula HOOC(CH2)4 COOH
MDL Number MFCD00004420
Melting Point 151°C to 154°C
Boiling Point 265°C (100 mmHg)
Synonym Hexanedioic acid
Quantity 100 mg
Formula Weight 146.14
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC