missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Adenosine, 99+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Absorbance | 0.76 to 0.80 (A250/A260),0.14 to 0.16 (A280/A260) |
| Melting Point | 233.0°C to 238.0°C |
| Color | White |
| Loss on Drying | 0.5% (105°C, 2 hrs) max. |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 100 g |
| Beilstein | 26, V,16, 335 |
| Merck Index | 15141 |
| Show More |
Safety and Handling
| EINECSNumber | 200-389-9 |
RUO – Research Use Only