missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenosine, 99+%

Catalog No. AC164045000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164045000 500 g Glass bottle
AC164040050 5 g Glass bottle
AC164040250 25 g Glass bottle
AC164041000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC164045000 Supplier Thermo Scientific Chemicals Supplier No. 164045000
Only null left

CAS: 58-61-7 | C10H13N5O4 | 267.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58-61-7
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00005752
InChI Key OIRDTQYFTABQOQ-KQYNXXCUSA-N
Synonym adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol
PubChem CID 60961
ChEBI CHEBI:16335
IUPAC Name (2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Absorbance 0.76 to 0.80 (A250/A260),0.14 to 0.16 (A280/A260)
Melting Point 233.0°C to 238.0°C
Color White
Loss on Drying 0.5% (105°C, 2 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 500 g
Beilstein 26, V,16, 335
Merck Index 15141
Specific Rotation − 60.00
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Spectrophotometry 98.0 to 102% (lambda max. 259nm,pH 7) molar absorptivity 15400+/-2% ,on dry basis
Transmittance 98% (c=0.5, H2O, 430nm) 1cm cell min.
Formula Weight 267.23
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Adenosine
Show More Show Less
EINECSNumber 200-389-9

RUO – Research Use Only