missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenosine, 99%

Catalog No. AAA1078109
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1078109 10 g
AAA1078118 50 g
AAA1078130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1078109 Supplier Thermo Scientific Chemicals Supplier No. A1078109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 58-61-7 | C10H13N5O4 | 267.25 g/mol

Adenosine serves as a nucleotide. It also plays an important role in the regulation of blood flow to various organs through vasodilation. It is actively involved in biochemical processes, such as energy transfer as adenosine triphosphate and adenosine diphosphate. Further, it is used in medication specifically as an antiarrhythmic agent to treat a number of supraventricular tachycardia. It is used to promote thickening of hair for people with thinning hair.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine serves as a nucleotide. It also plays an important role in the regulation of blood flow to various organs through vasodilation. It is actively involved in biochemical processes, such as energy transfer as adenosine triphosphate and adenosine diphosphate. Further, it is used in medication specifically as an antiarrhythmic agent to treat a number of supraventricular tachycardia. It is used to promote thickening of hair for people with thinning hair.

Solubility
Soluble in water, ammonium hydroxide and dimethyl sulfoxide. Insoluble in ethanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 58-61-7
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00005752
InChI Key OIRDTQYFTABQOQ-KQYNXXCUSA-N
Synonym adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol
PubChem CID 60961
ChEBI CHEBI:16335
IUPAC Name (2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Melting Point 234°C to 237°C
Odor Odorless
Assay Percent Range 99%
Quantity 10 g
Beilstein 93029
Merck Index 14,153
Solubility Information Soluble in water,ammonium hydroxide and dimethyl sulfoxide. Insoluble in ethanol.
Optical Rotation −70° (c=2 in 5% NaOH)
Formula Weight 267.25
Percent Purity 99%
Chemical Name or Material Adenosine
Show More Show Less
EINECSNumber 200-389-9
RTECSNumber AU7175000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only