missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-monophosphate disodium salt

Catalog No. AAJ6164314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6164314 25 g
AAJ6164306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6164314 Supplier Thermo Scientific Chemicals Supplier No. J6164314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4578-31-8 | C10H12N5Na2O7P | 391.19 g/mol

Adenosine-5'-monophosphate disodium salt is a nucleotide involved in the reactions of cellular energy transfers. It is useful for skin rejuvenation in dermatology and aesthetic dermatology. It acts as an activator of a class of protein kinases called as AMP-activated protein kinase (AMPK). Further, it is used as a substrate for enzymes such as AMP-thymidine kinase, AMP deaminase and 5'-nucleotidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine-5′-monophosphate disodium salt is a nucleotide involved in the reactions of cellular energy transfers. It is useful for skin rejuvenation in dermatology and aesthetic dermatology. It acts as an activator of a class of protein kinases called as AMP-activated protein kinase (AMPK). Further, it is used as a substrate for enzymes such as AMP-thymidine kinase, AMP deaminase and 5′-nucleotidase.

Solubility
Soluble in water, dimethyl sulfoxide and methanol.

Notes
Air and moisture sensitive. Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 4578-31-8
Molecular Formula C10H12N5Na2O7P
Molecular Weight (g/mol) 391.19
MDL Number MFCD00065023
InChI Key QGXLVXZRPRRCRP-IDIVVRGQSA-L
Synonym adenosine 5'-monophosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate, disodium amp, amp disodium salt, disodium 5'-amp, adenosine phosphate disodium, 5'-amp disodium salt, 5'-adenylic acid, disodium salt, unii-t1wz11dsrn, adenylic acid disodium salt
PubChem CID 20712
ChEBI CHEBI:81690
IUPAC Name disodium;[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Quantity 25 g
Beilstein 3586732
Merck Index 14,158
Solubility Information Soluble in water,dimethyl sulfoxide and methanol.
Formula Weight 391.2
Physical Form Powder
Chemical Name or Material Adenosine-5'-monophosphate disodium salt
EINECSNumber 224-961-2
RTECSNumber AU7481600
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only