missing translation for 'onlineSavingsMsg'
Learn More

Adenosine 3',5'-Cyclic Monophosphate Sodium Salt Hydrate 98.0+%, TCI America™

Catalog No. p-7117737
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2112100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2112100MG Supplier TCI America Supplier No. A2112100MG

Chemical Identifiers

CAS 37839-81-9
Molecular Formula C10H11N5NaO6P
Molecular Weight (g/mol) 351.19
MDL Number MFCD00069736
InChI Key BXJBFCKTIWRKMQ-GWKNMROSNA-M
Synonym adenosine 3',5'-cyclic monophosphate sodium salt monohydrate, c10h11n5o6p.na.h2o, sodium 3',5'-cyclic amp anion hydrate, adenosine 3',5'-cyclic monophosphate monosodium salt
PubChem CID 91872444
IUPAC Name sodium (4aR,6R,7R,7aS)-6-(6-amino-9H-purin-9-yl)-7-hydroxy-2-oxo-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate
SMILES [Na+].NC1=C2N=CN([C@@H]3O[C@@H]4COP([O-])(=O)O[C@H]4[C@H]3O)C2=NC=N1

Specifications

Color White
Quantity 100 mg
Formula Weight 351.19
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Adenosine 3′,5′-Cyclic Monophosphate Sodium Salt Hydrate
RTECSNumber AU7357900
TSCA No