missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-2',3'-cyclic monophosphate sodium salt, 97%

Catalog No. AAJ60558MD
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60558MD 250 mg
AAJ60558MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60558MD Supplier Thermo Scientific Chemicals Supplier No. J60558MD

CAS: 37063-35-7 | C10H11N5NaO6P | 351.19 g/mol

Adenosine-2’,3’-cyclic Monophosphate is an Adenosine (A280400) derivative useful as anticancer agent. It is also used as a Pharmaceutical intermediate, pharmaceutical research and chemical analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine-2’,3’-cyclic Monophosphate is an Adenosine (A280400) derivative useful as anticancer agent. It is also used as a Pharmaceutical intermediate, pharmaceutical research and chemical analysis.

Solubility
It is soluble in DMSO, water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Keep away fom strong oxidizing agents.

Chemical Identifiers

CAS 37063-35-7
Molecular Formula C10H11N5NaO6P
Molecular Weight (g/mol) 351.19
MDL Number MFCD00005757
InChI Key VSDSIACSNXHGOV-GWKNMROSNA-M
Synonym adenosine-2 inverted exclamation mark ,3 inverted exclamation mark-cyclic monophosphate sodium salt
PubChem CID 131698406
IUPAC Name [(3aR,4R,6R,6aR)-4-(6-aminopurin-9-yl)-2-oxido-2-oxo-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3,2]dioxaphosphol-6-yl]methanol;sodium
SMILES [Na+].NC1=C2N=CN([C@@H]3O[C@H](CO)[C@H]4OP([O-])(=O)O[C@@H]34)C2=NC=N1

Specifications

Melting Point 241°C to 243°C
Color White
Quantity 250 mg
Solubility Information It is soluble in DMSO,water.
Formula Weight 351.19
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Adenosine-2',3'-cyclic monophosphate sodium salt
EINECSNumber 253-328-3
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only