accessibility menu, dialog, popup

UserName

N(R)N(R)O(R)(R)NNNH2HOOPOOONa

CAS RN 37063-35-7

IUPAC Name: sodium (3aR,4R,6R,6aR)-4-(6-amino-9H-purin-9-yl)-6-(hydroxymethyl)-2-oxo-tetrahydro-2H-2λ⁵-furo[3,4-d][1,3,2]dioxaphosphol-2-olate
Synonyms: sodium 2',3'-cyclic amp
Molecular Weight (g/mol): 351.19
Molecular Formula: C10H11N5NaO6P
InChi Key: VSDSIACSNXHGOV-GWKNMROSNA-M
SMILES: [Na+].NC1=C2N=CN([C@@H]3O[C@H](CO)[C@H]4OP([O-])(=O)O[C@@H]34)C2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results