missing translation for 'onlineSavingsMsg'
Learn More

Adenine Sulfate Dihydrate 98.0+%, TCI America™

Catalog No. A015125G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A015125G 25 g
A0151100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A015125G Supplier TCI America Supplier No. A015125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6509-19-9
Molecular Formula C10H16N10O6S
Molecular Weight (g/mol) 404.36
MDL Number MFCD00150149
InChI Key MKPCNMXYTMQZBE-UHFFFAOYSA-N
Synonym 9h-purin-6-amine hemisulfate dihydrate, adenine hemisulfate dihydrate, 6-aminopurine sulfate dihydrate, bis ade sulfuric acid dihydrate, sulfuric acid bis vitamin b4 dihydrate, bis adenine sulfuric acid dihydrate, sulfuric acid-7h-purin-6-amine-water 1/2/2
PubChem CID 44134512
IUPAC Name bis(7H-purin-6-amine) sulfuric acid dihydrate
SMILES O.O.OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 368.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Adenine Sulfate Dihydrate
EINECSNumber (9)-0045
RTECSNumber AU7140000
TSCA Yes