accessibility menu, dialog, popup

UserName

CAS RN 6509-19-9

IUPAC Name: bis(7H-purin-6-amine) sulfuric acid dihydrate
Synonyms: bis(adenine) sulfuric acid dihydrate
Molecular Weight (g/mol): 404.36
Molecular Formula: C10H16N10O6S
InChi Key: MKPCNMXYTMQZBE-UHFFFAOYSA-N
SMILES: O.O.OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)