missing translation for 'onlineSavingsMsg'
Learn More

Adenine Sulfate Dihydrate, >97%

Catalog No. aaj1045022
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
Packaging:
Plastic bottle
Catalog No. Quantity Packaging
AAJ1045022 100 g Plastic bottle
1 options

Catalog No. AAJ1045022

Supplier: Thermo Fisher Scientific J1045022

Only null left

CAS: 321-30-2 | C10H12N10O4S | 368.33 g/mol

 

Chemical Identifiers

CAS 321-30-2
Molecular Formula C10H12N10O4S
Molecular Weight (g/mol) 368.33
MDL Number MFCD00213655
InChI Key LQXHSCOPYJCOMD-UHFFFAOYSA-N
Synonym adenine sulfate, adenine hemisulfate, 7 h-purin-6-amine sulfate 2:1, adeninsulfat, adeninium sulfate, adenine sulfate 2:1, adeninsulfat german, 1h-purin-6-amine, sulfate 2:1, diadenine sulphate, unii-741gjf3k9m
PubChem CID 9449
IUPAC Name 7 H-purin-6-amine;sulfuric acid
SMILES OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1

Specifications

Melting Point ∼210°C (decomposition)
Packaging Plastic bottle
Quantity 100 g
Beilstein 3820263
Formula Weight 404.36
Percent Purity ≥97%
Physical Form Crystalline Powder
Chemical Name or Material Adenine Sulfate Dihydrate
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 206-286-5
RTECSNumber AU7140000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only