missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenine sulfate, 98%, synthetic

Catalog No. p-50837
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163630250 25 g Plastic bottle
AC163631000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC163630250 Supplier Thermo Scientific Chemicals Supplier No. 163630250
Only null left

CAS: 321-30-2 | C10H12N10O4S | 368.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 321-30-2
Molecular Formula C10H12N10O4S
Molecular Weight (g/mol) 368.33
MDL Number MFCD00213655
InChI Key LQXHSCOPYJCOMD-UHFFFAOYSA-N
Synonym adenine sulfate, adenine hemisulfate, 7h-purin-6-amine sulfate 2:1, adeninsulfat, adeninium sulfate, adenine sulfate 2:1, adeninsulfat german, 1h-purin-6-amine, sulfate 2:1, diadenine sulphate, unii-741gjf3k9m
PubChem CID 9449
IUPAC Name 7H-purin-6-amine;sulfuric acid
SMILES OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1

Specifications

Absorbance 0.74 to 0.78 (A250/A260), 0.34 to 0.38 (A280/A260)
CAS Min % 97.5
CAS Max % 100.0
Melting Point 285.0°C
Color White to Yellow
Loss on Drying 2% max. (1 g, 105°C, 5 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 25 g
Beilstein 26, V,16, 100
Solubility Information Solubility in water: soluble
Spectrophotometry 97.5% min. (lambda max. 262nm,pH 1) molar absorptivity 13100+/-2%,on dry basis
Transmittance 95% min. (c=0.5, H2O, 430nm) 1cm cell
Formula Weight 184.17
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Adenine sulfate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 206-286-5
RTECSNumber AU7140000
TSCA TSCA

RUO – Research Use Only