Learn More
Thermo Scientific Chemicals Adenine sulfate, 98+%
CAS: 321-30-2 | C10H12N10O4S | 368.33 g/mol
Supplier: Thermo Scientific Chemicals A1696409
Description
It has a wide range of chemical and biochemical roles employing as a regulatory molecule. It acts as a precursor, substrate, or cofactor in diverse biochemical pathways.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt has a wide range of chemical and biochemical roles employing as a regulatory molecule. It acts as a precursor, substrate, or cofactor in diverse biochemical pathways.
Solubility
Soluble in water (4 mg/ml), ethanol (<1mg/mL at 25°C), DMSO (<1mg/mL at 25°C), and HCl (10 mg/ml).
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Specifications
| Adenine sulfate | |
| ∽210°C (decomposition) | |
| C10H12N10O4S | |
| 10 g | |
| Hygroscopic | |
| adenine sulfate, adenine hemisulfate, 7h-purin-6-amine sulfate 2:1, adeninsulfat, adeninium sulfate, adenine sulfate 2:1, adeninsulfat german, 1h-purin-6-amine, sulfate 2:1, diadenine sulphate, unii-741gjf3k9m | |
| LQXHSCOPYJCOMD-UHFFFAOYSA-N | |
| 7H-purin-6-amine;sulfuric acid | |
| 9449 | |
| ≥98% |
| 321-30-2 | |
| >98% | |
| MFCD00213655 | |
| 3820263 | |
| 14,152 | |
| Soluble in water (4mg/ml),ethanol (<1mg/ml at 25°C),DMSO (<1mg/ml at 25°C),and HCl (10mg/ml). | |
| OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1 | |
| 368.33 | |
| 368.34 |
Chemical Identifiers
| 321-30-2 | |
| 368.33 | |
| LQXHSCOPYJCOMD-UHFFFAOYSA-N | |
| 9449 | |
| OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1 |
| C10H12N10O4S | |
| MFCD00213655 | |
| adenine sulfate, adenine hemisulfate, 7h-purin-6-amine sulfate 2:1, adeninsulfat, adeninium sulfate, adenine sulfate 2:1, adeninsulfat german, 1h-purin-6-amine, sulfate 2:1, diadenine sulphate, unii-741gjf3k9m | |
| 7H-purin-6-amine;sulfuric acid |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 206-286-5
RTECSNumber : AU7140000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only